
2-Diethoxyphosphorylethylazanium 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:203192-98-7
:
Formula:C₆H₁₆NO₃P·C₂H₂O₄
MDL Number:MFCD00800256
:271.21 g/mol
Melting Pt:135…141 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:135 to 141 °C
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together