
Diethyl (aminomethyl)phosphonate oxalate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:117196-73-3
:
Formula:C₇H₁₆NO₇P
MDL Number:MFCD00800255
:257.179 g/mol
Melting Pt:118…126 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:118 to 126 °C
- Titration with HClO4:≥94 %
Specification Test Results
Frequently Bought Together