Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
 
2-Chloro-2-oxo-1,3,2-dioxaphospholane 95%
 
undefined
 
2-Chloro-2-oxo-1,3,2-dioxaphospholane 95%

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4475 to 1.4515 (20°C, 589 nm)
  • Titration Argentometric:
    ≥94 % (ex Cl)

 

 

Frequently Bought Together