
Diethyl (chloromethyl)phosphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3167-63-3
:
Formula:C₅H₁₂ClO₃P
MDL Number:MFCD00010189
:186.57 g/mol
Boiling Pt:109…110 °C (10 mmHg)
Density:1.2 g/cm³
Flash Pt:86 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4400 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together