Order Entry
Czech Republic
ContactUsLinkComponent
Acrylamide : Bisacrylamide (37.5:1) 40% mix in water for biochemistry
Irritant
Health
Acrylamide : Bisacrylamide (37.5:1) 40% mix in water for biochemistry
Certificates
undefined
Irritant
Health
Acrylamide : Bisacrylamide (37.5:1) 40% mix in water for biochemistry

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • UV:
    (1 % in water)
  • Acrylic acid (HPLC):
    ≤0.001 %
  • Conductivity:
    ≤3 µS/cm

Specification Test Results

Related Information

Frequently Bought Together