

Acrylamide : Bisacrylamide (29:1) 40% mix in water for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-06-1
MDL Number:MFCD00080848
Density:1.035 g/cm³
Storage Temperature:Refrigerator
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless Solution
- UV:(1 % in water)
- Acrylic acid (HPLC):≤0.001 %
- Conductivity:≤3 µS/cm
Specification Test Results
Frequently Bought Together