Benzofuran-2-carboxaldehyde 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4265-16-1
:
Formula:C₉H₆O₂
MDL Number:MFCD00015463
:146.145 g/mol
Density:1.206 g/cm³
Flash Pt:>110 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6300 to 1.6340 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
Frequently Bought Together