Order Entry
Czech Republic
ContactUsLinkComponent
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one 98%
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one 98%
Certificates
undefined
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one 98%

Specification Test Results

  • Appearance:
    Off-white to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    160 to 170 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together