Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51298-62-5
:
Formula:C₇H₁₅N₅O₄
MDL Number:MFCD00039052
:269.69 g/mol
Melting Pt:159…161 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline solid
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:+14.0° to +16.0° (24°C, 589 nm) (c=3, CH3OH)
Specification Test Results
Frequently Bought Together