Order Entry
Czech Republic
ContactUsLinkComponent
Nω-Methyl-L-arginine monoacetate 99%
Nω-Methyl-L-arginine monoacetate 99%
Certificates
undefined
Nω-Methyl-L-arginine monoacetate 99%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    180 to 190 °C
  • TLC:
    ≥98.5 %
  • Specific optical rotation:
    +9° to +13° (20 °C, 589 nm) (c=4, water)

Specification Test Results

Related Information

Frequently Bought Together