
2,2'-Bipyridyl-1,1'-dioxide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7275-43-6
:
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD00065169
:188.19 g/mol
Melting Pt:296…298 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to cream Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together