
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%
:126456-43-7
:
:C₉H₁₁NO
:MFCD00216655
:149.19 g/mol
:117…121 °C
:GHS07
:Ambient
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Melting point:117 to 121 °C
- GC:≥98.5 %
- Specific optical rotation:-41.5° to -46.5° (20 °C, 589 nm) (c=1, CH3OH)
Frequently Bought Together