ContactUsLinkComponent
Ethoxyacetylene 40% (w/w) in hexanes
Irritant
Health
Environment
Flammable
Ethoxyacetylene 40% (w/w) in hexanes
Certificates
undefined
Irritant
Health
Environment
Flammable
Ethoxyacetylene 40% (w/w) in hexanes
Specification Test Results
  • Appearance:
    Clear orange to brown Solution
  • Identification:
    confirmed by 1H-NMR
  • NMR (1H-NMR):
    ca. 40 % w/w (Typical 35 to 55 % w/w)