

Dodecanedioyl dichloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4834-98-4
:
Formula:C₁₂H₂₀Cl₂O₂
MDL Number:MFCD00012243
:267.19 g/mol
Boiling Pt:140 °C (.5 mmHg)
Density:1.06 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear yellow to brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4670 to 1.4690 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together