Order Entry
Czech Republic
ContactUsLinkComponent
Ethephon(2-chloroethylphosphonic acid) 90% tech.
Corrosive
Toxic
Environment
Ethephon(2-chloroethylphosphonic acid) 90% tech.
Catalog # ACRO301630050
CAS Number:  
undefined
Corrosive
Toxic
Environment
Ethephon(2-chloroethylphosphonic acid) 90% tech.
Catalog # ACRO301630050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    65 to 75 °C
  • Titration with NaOH:
    ≥88 %

Specification Test Results

About this item

CAS Number:16672-87-0
 :
Formula:C₂H₆ClO₃P
MDL Number:MFCD00002142
Melting Pt:74…75 °C
Storage Temperature:Ambient
UN:2928
ADR:6.1,II
 :GHS05
 :GHS06
 :GHS09