Order Entry
Czech Republic
ContactUsLinkComponent
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)
Irritant
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)
Certificates
undefined
Irritant
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)

Specification Test Results

  • Appearance:
    Beige to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    80 to 84 °C
  • NMR:
    ≥94 %

Specification Test Results

Related Information

Frequently Bought Together