

Chloromethyl butyrate 99%
:33657-49-7
:
:C₅H₉ClO₂
:MFCD00216961
:136.58 g/mol
:150 °C (993mbar)
:1,074 g/cm³
:55 °C
:Ambient
:2924
:3,III
:GHS05
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4220 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together