

(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 99%
Catalog # ACRO294662500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:48 to 51 °C
- Specific optical rotation:+48° to +51° (20 °C, 589 nm) (c=2, MeOH)
- Titration with HClO4:98.5 to 101.5 %
Specification Test Results
About this item
:
:240.35 g/mol
Melting Pt:48…51 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS07
:GHS05
CAS Number:118628-68-5
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00216955