Order Entry
Czech Republic
ContactUsLinkComponent
4-Epichlortetracycline hydrochloride, 97%, can be used as secondary standard
4-Epichlortetracycline hydrochloride, 97%, can be used as secondary standard
Certificates
undefined
4-Epichlortetracycline hydrochloride, 97%, can be used as secondary standard
Certificates

Specifications

  • CAS No.:
    101342-45-4
  • Melting point:
    250°C (D)
  • Formula:
    C₂₂H₂₃ClN₂O₈.ClH
  • Description:
    4-Epichlortetracycline hydrochloride

Specifications

Related Information

Frequently Bought Together