Dimethyl (+)-2,3-O-isopropylidene-D-tartrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37031-30-4
:
Formula:C₉H₁₄O₆
MDL Number:MFCD00040504
:218.21 g/mol
Boiling Pt:139 °C (21hPa)
Density:1.15 g/cm³
Flash Pt:135 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4386 to 1.4406 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:+52° to +56° (20°C, 589 nm) (neat)
Specification Test Results
Frequently Bought Together