ContactUsLinkComponent
3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water
Irritant
Corrosive
Flammable
Environment
3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water
Certificates
undefined
Irritant
Corrosive
Flammable
Environment
3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water
Certificates
Specification Test Results
  • Appearance:
    White Moist powder
  • Titration Iodometric:
    70 to 75 %
  • Water:
    % balance (K.F.)
  • HPLC:
    ≤0.2 % di-m-Chlorobenzoyl peroxide
  • 3-Chlorobenzoic acid:
    ≤12 %