ContactUsLinkComponent
Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal®
Irritant
Corrosive
Flammable
Health
Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal®
Certificates
undefined
Irritant
Corrosive
Flammable
Health
Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal®
Specification Test Results
  • Appearance:
    Dark brown Solution
  • Titration with HCl:
    24 to 26 %