
2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21306-21-8
:
Formula:C₆Cl₆O
MDL Number:MFCD00019431
:300.765 g/mol
Melting Pt:46…49 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Crystalline solid
- Infrared spectrum:Conforms
- Melting point:46 to 52 °C
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together