Order Entry
Czech Republic
ContactUsLinkComponent
1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide 98%
Irritant
1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide 98%
Catalog # ACRO235060010
CAS Number:  
undefined
Irritant
1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide 98%
Catalog # ACRO235060010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Beige to grey-green to brown Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 %

Specification Test Results

About this item

 :
 :246.104 g/mol
Melting Pt:210 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:52768-23-7
Formula:C₉H₁₂BrNO₂
MDL Number:MFCD00078115