4-Epianhydrotetracycline hydrochloride, 97%, can be used as a secondary standard
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 4465-65-0
MDL No.: MFCD00151455
GHS Symbols: GHS07, GHS08,
Melting Point: 220 °C (D)
Appearance: Orange Powder
Storage Temperature: Ambient
Signal word: Warning
Specifications
- CAS No.:4465-65-0
- Formula:C₂₂H₂₂N₂O₇.HCl
- Description:4-Epianhydrotetracycline hydrochloride
Specifications
Frequently Bought Together