ContactUsLinkComponent
Atropine sulphate monohydrate 97+%
Toxic
Atropine sulphate monohydrate 97+%
Certificates
undefined
Toxic
Atropine sulphate monohydrate 97+%
Certificates
Specification Test Results
  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Water:
    2.0 to 4.0 % (K.F.)
  • Residue after ignition:
    ≤0.2 %
  • Specific optical rotation:
    -0.50° to +0.05° (25°C, 589 nm) (c=10, water) (on anhydrous basis)
  • Titration with HClO4:
    97.0 to 103.0 % (On anhydrous basis)