Graduated pipettes, with safety bulb, Qualicolor, Colour-Coded
Supplier: VTR Glass
Certificates
About this item
Sodium-potassium glass, class B.
- Calibrated for discharge, whole volume
- Cannot be autoclaved at 121 °C
Certifications: DIN 12696, ISO 835
Frequently Bought Together