0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Pumps
Drum/Transfer Pumps
Accessories for drum pumps, Asecos
Accessories for drum pumps, Asecos
MSGA2790
:
asecos
Accessories for drum pumps, Asecos
MSGA2790
:
asecos
Description:
Antistatic kit, includes 3 copper cables of different colours with clips, supplied in a plastic case
Pk:
1