Anti-IgG Llama Polyclonal Antibody (CF™ 488A)
20454-1MG
: Biotium
- Antibody type:Secondary
- Antigen name:Mouse IgG
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 488A
- Host:Llama
- Reactivity:Mouse
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, 15 mg/ml BSA, 20 mg/mL trehalose (after reconstitution)
- Storage temperature:Store at –20 °C.
- Concentration:2 mg/ml (after reconstitution)
- Purification:Affinity purified
- Size:1 mg
- Pk:1 mg
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™488A (Ex/Em 490/515 nm) is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF™488A has brightness and photostability comparable to Alexa Fluor® 488.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse