Anti-IgG Llama Polyclonal Antibody (CF™ 647)
Catalog # 20453-1MG
Supplier: Biotium
Specifications
- Antibody type:Secondary
- Antigen name:Rabbit IgG
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 647
- Host:Llama
- Reactivity:Rabbit
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, 15 mg/ml BSA, 20 mg/mL trehalose (after reconstitution)
- Storage temperature:Store at –20 °C.
- Concentration:2 mg/ml (after reconstitution)
- Purification:Affinity purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Llama anti-rabbit CF™ dye conjugates are prepared by labeling affinity purified llama anti-rabbit IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Llama
Isotype:
Reactivity: Rabbit