Anti-IgG Llama Polyclonal Antibody (CF™ 568)
Dodavatel: Biotium
Certifikáty
O tomto produktu
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. Red fluorescent CF™568 (Ex/Em 562/583 nm) produces much brighter antibody conjugates than Alexa Fluor® 568, and has superior photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF568
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Specifikace
- Catalog No:
- 20455-50UL
- 20455-500UL
- Symbol antigenu:
- IgG
- IgG
- Název antigenu:
- Mouse IgG
- Mouse IgG
- Druh protilátky:
- Sekundární
- Sekundární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- CF® 568
- CF® 568
- Reaktivita:
- Mouse
- Mouse
- Hostitel:
- Llama
- Llama
- Formulář:
- Liquid
- Liquid
- Čištění:
- Affinity purified
- Affinity purified
- Koncentrace:
- 2 mg/mL
- 2 mg/mL
- Křížová adsorbce:
- No
- No
- Skladovací pufr:
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Skladovací teplota:
- Store at –20 °C.
- Store at –20 °C.
Specifikace
Frequently Bought Together