About this item
Llama anti-rabbit CF™ dye conjugates are prepared by labeling affinity purified llama anti-rabbit IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Llama
Isotype:
Reactivity: Rabbit
Specifications
- Catalog No:
- 20452-50UL
- 20452-500UL
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Rabbit IgG
- Rabbit IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Llama
- Llama
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Concentration:
- 2 mg/mL
- 2 mg/mL
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Storage temperature:
- Store at –20 °C.
- Store at –20 °C.