Specifications
- Antibody type:Tag
- Antigen name:Myc Tag (EQKLISEEDL)
- Antigen symbol:Myc Tag
- Clonality:Polyclonal
- Conjugation:CF® 594
- Host:Rabbit
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Storage temperature:Store at –20 °C.
- Concentration:1 mg/mL
- Purification:Affinity purified
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.
Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.
Type: Tag
Antigen: Myc Tag
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype:
Reactivity: MYC tag