Bulb pipettes, with safety bulb, QUALICOLOR, COLOR CODE, two marks, Class B
Supplier: VTR Glass
Certificates
About this item
Soda-lime glass.
- Enlarged form
- Not autoclavable at 121 °C
Certifications: DIN 12690, ISO 648
Frequently Bought Together
Soda-lime glass.