Bulb pipettes, with safety bulb, Qualicolor, colour-coded, one mark, Class B
Supplier: VTR Glass
Certificates
About this item
Soda-lime glass.
- Enlarged form
- Not autoclavable at 121 °C
Certifications: DIN 12690, ISO 648
Frequently Bought Together
Soda-lime glass.