SDS precipitation kit, SDS-Out™
Catalog # PIER20308
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:SDS-Out™ SDS precipitation kit, sufficient for precipitation of SDS from 200 ml of protein solution
- Environmentally Preferable:
Specifications
About this item
SDS-Out™ SDS Precipitation Kit provides convenient removal of SDS from small-volume (0,1 to 10 ml) protein samples.
- Convenient removal of SDS from microliter-volume samples
- Used for samples containing 0,1 to 1% SDS
- Minimal dilution of small-volume samples
- Protein recovery is 90 to 100%
Delivery: The kit includes SDS precipitation reagent (10 ml), spin cup columns (12 columns) and collection tubes (12 tubes).