Order Entry
Czech Republic
ContactUsLinkComponent
Oxalic acid 6,3% in aqueous solution USP Test Solution (TS)
Oxalic acid 6,3% in aqueous solution USP Test Solution (TS)
Catalog # 85358.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxalic acid 6,3% in aqueous solution USP Test Solution (TS)
Catalog # 85358.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :90.04 g/mol
Merck Index:13,06980
Melting Pt:99 to 100 °C
Storage Temperature:Ambient
EINECS:0
UN:3265
ADR:8,III
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573