ContactUsLinkComponent
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent
Catalog # PIER23238
CAS Number:  
undefined
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent
Catalog # PIER23238
CAS Number:  
Specifications
  • Pk:
    300 mL
  • Description:
    Coomassie plus (bradford) reagent, sufficient reagent to perform 200 standard assays or 1000 microplate assays
  • Seq:
    01
Specifications
About this item
The bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration. The Pierce™ coomassie plus protein assay is a ready-to-use, reducing agent-compatible, improved bradford assay reagent to quickly measure (A595 nm) total protein concentration compared to a protein standard.
  • Sensitive, fast and consistent protein assay kit
  • Determine protein concentrations from 1 to 1500 μg/ml
  • Ready-to-use dye binding reagent formulation
  • Convenient microplate or cuvette format
  • Compatible with reducing sugars, reducing substances and thiols
This modification of the well-known bradford method greatly reduces the tendency of coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, coomassie plus reagent results in significantly less protein-to-protein variation than is observed with other bradford-type coomassie formulations.