ContactUsLinkComponent
Cylinders for urinometers
Cylinders for urinometers
Catalog # 612-2764
undefined
Cylinders for urinometers
Catalog # 612-2764
Specifications
  • Ø:
    28 mm
  • Height:
    175 mm
  • Seq:
    901
  • Pk:
    1
Specifications
About this item
Jar, made of glass.
With base and spout for urinometers.