Subcellular protein fractionation kit for tissues
Supplier: Thermo Fisher Scientific
Certificates
About this item
This kit is ideal for cell fractionation and enrichment of proteins from five different cellular compartments within two hours.
- Optimized - formulations and protocols specific for fractionation of tissue
- Complete - obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
- Complete - obtain protein fractions from a single kit
- Convenient - unique tissue strainer allows rapid removal of tissue debris
- Versatile - extract proteins from a variety of tissue types, including liver, heart, brain, kidney, lung and spleen
- Compatible - use extracts for downstream applications such as protein assays, Western blotting, protein assays, electrophoretic mobility shift assays, reporter-gene and enzyme-activity assays
Delivery: Kit includes four extraction buffers, tissue strainers, a stabilized nuclease and halt protease inhibitor cocktail.
Frequently Bought Together