Pycnometer acc. to Hubbard
Catalog # VTRB632431762023
Supplier: Kavalier
Specifications
- Description:Pycnometer, 25 ml
- Pk:1
Specifications
About this item
SIMAX® borosilicate glass
For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses.