2-Iminothiolane hydrochloride (Traut's Reagent), Pierce™
Catalog # PIER26101
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:137.625 g/mol
Storage Temperature:Refrigerator
CAS Number:4781-83-3
Formula:C₄H₈ClNS
MDL Number:MFCD00039013