Order Entry
Czech Republic
ContactUsLinkComponent
Pierce™ DNA Extraction Reagent Kit, Yeast
Pierce™ DNA Extraction Reagent Kit, Yeast
Catalog # PIER78870
CAS Number:  
undefined
Pierce™ DNA Extraction Reagent Kit, Yeast
Catalog # PIER78870
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    Yeast DNA extraction reagent kit, sufficient for 50 purifications from 10 ml cultures
  • Description:
    Yeast DNA Extraction Reagent Kit

Specifications

About this item

The Pierce™ yeast DNA extraction reagent kit uses Y-PER yeast protein extraction reagent to quickly extract and purify genomic and plasmid DNA from yeast.

  • Greater yields than traditional methods
  • Eliminates need for glass beads or harsh enzyme treatments
  • Yields DNA that is compatible with PCR amplification
  • Scalable to allow purification from single colonies up to 500 ml of cultured yeast
Delivery: Kit includes Y-PER yeast protein extraction reagent (25 ml), DNA releasing Agent A and B (20 ml of each), and protein removal reagent (10 ml).