Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
SANTSC-2354
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Storage temperature:+4 °C
- Size:200 µg/0,5 ml
- Pk:0,5 mL
Western blotting of immunoglobulin-rich tissues and cells at a dilution of 1:500 - 1:10000. Starting dilution has to be 1:2000.
It is pre-adsorbed with human IgG.
Type: Secondary
Antigen:
Clonality:
Clone:
Conjugation: Horseradish peroxidase
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse