ContactUsLinkComponent
Ultrapure lyophilisation stopper, WHEATON®
Ultrapure lyophilisation stopper, WHEATON®
Catalog # WHEAW224100-407
Supplier:  DWK Life Sciences
undefined
Ultrapure lyophilisation stopper, WHEATON®
Catalog # WHEAW224100-407
Supplier:  DWK Life Sciences
Specifications
  • Ø bottom:
    20,0 mm
  • Description:
    Stopper with 3 legs
  • Pk:
    1.000
Specifications
About this item
This ultrapure lyophilization stopper is a very low moisture adsortion stopper. Manufactured from an ultra pure bromobutyl formulation with extremely high chemical purity and low gas permeability.
  • Very low level of adsortion of the preservatives present in aqueous parenteral solutions
  • Autoclavable
  • Free of latex