Specifications
- Pk:5 mg
- CAS No.:9001-78-9
- Source:Intestine
- Species:Calf
- Seq:01
- Storage conditions:0 °C
- Form:Lyophilized powder
- Enzyme name:Alkaline Phosphatase
- pH:9,8
Specifications
About this item
Alkaline phosphatase is a hydrolase enzyme responsible for removing phosphate groups from many types of molecules, including nucleotides, proteins, and alkaloids.
- Enzyme Commission Number: E.C. 3.1.3.1
- Isoelectric point (pI): 5,7 (Lit.)
It is used for removing phosphate monoester to prevent self ligation. It can be also be used as a label for enzyme immunoassays.
It is fully active in the presence of sodium azide (0,05%) and Triton X-100 (0,1%).
Activators: Zn, Mg²⁺, Ca²⁺, Tris buffer, diethanolamine
Inhibitors: Acidification to pH 4,5 reversibly inactives the enzyme; inorganic phosphate, chelating agents such as EDTA
Unit definition: One unit liberates one µmole of p-nitrophenol per minute at 37 °C, pH 10,15 in Tris buffer. The enzyme activity is determined by measuring the increase in optical density at 405 nm resulting from the hydrolysis of p-nitrophenyl phosphate.