Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acid blue 29
Acid blue 29, black powder C.I. 20460
Acid blue 29, black powder C.I. 20460
Acid blue 29, black powder C.I. 20460
ICNA0215259625
:
MP Biomedicals
:
Acid blue 29, black powder C.I. 20460
ICNA0215259625
:
MP Biomedicals
:
Pk:
25 g
Seq:
3308
Identity Test:
Passes
UV/Vis: λmax (water):
600 - 605 nm
:
:
616.50 g/mol
:
Ambient
:
5850-35-1
:
C₂₂H₁₄N₆O₉S₂Na₂
:
MFCD00009960