ContactUsLinkComponent
Nitrite standard solution
Nitrite standard solution
  HACH2340249
 :  Hach
 :  
undefined
Nitrite standard solution
  HACH2340249
 :  Hach
 :  
  • Concentration:
    250 mg/l as N
  • Pk:
    500 mL
  • Description:
    Nitrite standard solution, stock, 500 ml
For accuracy checks in nitrite determinations.