Order Entry
Czech Republic
ContactUsLinkComponent
 
Formaldehyde standard solution
Catalog # 705-2033
Supplier:  Hach
CAS Number:  
undefined
Formaldehyde standard solution
Catalog # 705-2033
Supplier:  Hach
CAS Number:  

Specifications

  • Concentration:
    4000 mg/l CH₂O
  • Pk:
    16
  • CAS No.:
    50-00-0
  • Description:
    Formaldehyde standard solution, (NIST)
  • Formula:
    CH₂O
  • Description:
    Formaldehyde
  • Pk Info:
    16×10 ml Voluette ampoules

Specifications

About this item

For accuracy checks in formaldehyde determination.