Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Přihlásit se
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Control standard
Control standard
Control standard
Catalog # HACHLCW828
Supplier:
Hach
CAS Number:
Expand image
Control standard
Catalog # HACHLCW828
Supplier:
Hach
CAS Number:
Specifications
Concentration:
25 mg/l as NO3
Pk:
500 mL
Description:
Control standard for nitrate determination
Specifications
About this item
Product Details
Product Details